missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DDB1- and CUL4-associated factor 4 Protein
A DNA sequence encoding the Homo sapiens (Human) DDB1- and CUL4-associated factor 4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7992-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001156980.1 | |
DDB1- and CUL4-associated factor 4 Protein | |
Research Use Only | |
DCAF4 | |
Recombinant Protein | |
E. coli | |
MNKSRWQSRR RHGRRSHQQN PWFRLRDSED RSDSRAAQPA HDSGHGDDES PSTSSGTAGT SSVPELPGFY FDPEKKRYFR LLPGHNNCNP LTKESIRQKE MESKRLRLLQ EEDRRKKIAR MGFNASSMLR KSQLGFLNVT NYCHLAHELR LSCMERKKVQ IRSMDPSALA SDRFNLILAD TNSDRLFTVN DVKVGGSKYG IINLQSLKTP TLKVFMHENL YFTNRKVNSV CWASLNHLDS HILLCLMGLA ETPGCATLLP ASLFVNSHPG IDRPGMLCSF RIPGAWSCAW SLNIQANNCF STGLSRRVLL TNVVTGHRQS FGTNSDVLAQ QFALMAPLLF NGCRSGEIFA IDLRCGNQGK GWKATRLFHD SAVTSVRILQ DEQYLMASDM AGKIKLWDLR TTKCVRQYEG HVNEYAYLPL HVHEEEGILV AVGQDCYTRI WSLHDARLLR TIPSPYPASK ADIPSVAFSS RLGGSRGAPG LLMAVGQDLY CYSYS | |
Greater than 90% as determined by SDS-PAGE. |
26094 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |