missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DEFB1 / Beta-defensin 1 Protein
A DNA sequence encoding the Human DEFB1 / Beta-defensin 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10429-ec-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_005209.1 | |
DEFB1 / Beta-defensin 1 Protein | |
Research Use Only | |
DEFB1 | |
The biological activity determined by a chemotaxis bioassay using CD34+ dendritic cells is in a concentration range of 100.0-1000.0 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered 20 mM PBS, pH 7.4, 130 mM NaCl, lyophilized |
1672 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
GNFLTGLGHR SDHYNCVSSG GQCLYSACPI FTKIQGTCYR GKAKCCK | |
Greater than 98% as determined by SDS-PAGE and HPLC. |