missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DUSP13 Protein
A cDNA sequence encoding the DUSP13 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11711-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
DUSP13 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMDSLQK QDLRRPKIHG AVQASPYQPP TLASLQRLLW VRQAATLNHI DEVWPSLFLG DAYAARDKSK LIQLGITHVV NAAAGKFQVD TGAKFYRGMS LEYYGIEADD NPFFDLSVYF LPVARYIRAA LSVPQGRVLV HCAMGVSRSA TLVLAFLMIC ENMTLVEAIQ TVQAHRNICP NSGFLRQLQV LDNRLGRETG RF | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The DUSP13 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer, pH 8.0, 10% glycerol, 2mM DTT and 50mM NaCl. |