missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dynein light chain 1, cytoplasmic Protein
A DNA sequence encoding the Homo sapiens (Human) Dynein light chain 1, cytoplasmic, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8357-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001032583.1 | |
Dynein light chain 1, cytoplasmic Protein | |
Research Use Only | |
DYNLL1 | |
Recombinant Protein | |
E. coli | |
MCDRKAVIKN ADMSEEMQQD SVECATQALE KYNIEKDIAA HIKKEFDKKY NPTWHCIVGR NFGSYVTHET KHFIYFYLGQ VAILLFKSG | |
Greater than 90% as determined by SDS-PAGE. |
8655 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |