missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DYNLL2 Protein
A cDNA sequence encoding the DYNLL2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11718-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
DYNLL2 Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
DYNLL2 protein solution (1 mg/1ml) containing 20mM Tris-HCl buffer (pH 8.0), 30% glycerol, 0.2M NaCl and 1mM DTT. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSDRKAVIKN ADMSEDMQQD AVDCATQAME KYNIEKDIAA YIKKEFDKKY NPTWHCIVGR NFGSYVTHET KHFIYFYLGQ VAILLFKSG |