missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF2B1 Protein
A DNA sequence encoding the Homo sapiens (Human) EIF2B1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7612-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001405.1 | |
EIF2B1 Protein | |
Research Use Only | |
EIF2B1 | |
Recombinant Protein | |
E. coli | |
MDDKELIEYF KSQMKEDPDM ASAVAAIRTL LEFLKRDKGE TIQGLRANLT SAIETLCGVD SSVAVSSGGE LFLRFISLAS LEYSDYSKCK KIMIERGELF LRRISLSRNK IADLCHTFIK DGATILTHAY SRVVLRVLEA AVAAKKRFSV YVTESQPDLS GKKMAKALCH LNVPVTVVLD AAVGYIMEKA DLVIVGAEGV VENGGIINKI GTNQMAVCAK AQNKPFYVVA ESFKFVRLFP LNQQDVPDKF KYKADTLKVA QTGQDLKEEH PWVDYTAPSL ITLLFTDLGV LTPSAVSDEL IKLYL | |
Greater than 80% as determined by SDS-PAGE. |
1967 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |