missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Endostatin Protein
A cDNA sequence encoding the Endostatin was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10588-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Endostatin Protein | |
Research Use Only | |
The activity was found to be 16,000IU/mg. | |
Human | |
Untagged | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
The recombinant Human Endostatin is formulated with 30mM Sodium acetate buffer and Mannitol, pH 5.5. |