missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Epoxide hydrolase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Epoxide hydrolase 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5985-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000111.1 | |
Epoxide hydrolase 1 Protein | |
Research Use Only | |
EPHX1 | |
Recombinant Protein | |
E. coli | |
MWLEILLTSV LGFAIYWFIS RDKEETLPLE DGWWGPGTRS AAREDDSIRP FKVETSDEEI HDLHQRIDKF RFTPPLEDSC FHYGFNSNYL KKVISYWRNE FDWKKQVEIL NRYPHFKTKI EGLDIHFIHV KPPQLPAGHT PKPLLMVHGW PGSFYEFYKI IPLLTDPKNH GLSDEHVFEV ICPSIPGYGF SEASSKKGFN SVATARIFYK LMLRLGFQEF YIQGGDWGSL ICTNMAQLVP SHVKGLHLNM ALVLSNFSTL TLLLGQRFGR FLGLTERDVE LLYPVKEKVF YSLMRESGYM HIQCTKPDTV GSALNDSPVG LAAYILEKFS TWTNTEFRYL EDGGLERKFS LDDLLTNVML YWTTGTIISS QRFYKENLGQ GWMTQKHERM KVYVPTGFSA FPFELLHTPE KWVRFKYPKL ISYSYMVRGG HFAAFEEPEL LAQDIRKFLS VLERQ | |
Greater than 80% as determined by SDS-PAGE. |
2052 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |