missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ErbB3 Protein
A cDNA sequence encoding the ErbB3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11803-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
ErbB3 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
SEVGNSQAVC PGTLNGLSVT GDAENQYQTL YKLYERCEVV MGNLEIVLTG HNADLSFLQW IREVTGYVLV AMNEFSTLPL PNLRVVRGTQ VYDGKFAIFV MLNYNTNSSH ALRQLRLTQL TEILSGGVYI EKNDKLCHMD TIDWRDIVRD RDAEIVVKDN GRSCPPCHEV CKGRCWGPGS EDCQTLTKTI CAPQCNGHCF GPNPNQCCHD ECAGGCSGPQ DTDCFACRHF NDSGACVPRC PQPLVYNKLT FQLEPNPHTK YQYGGVCVAS CPHNFVVDQT SCVRACPPDK MEVDKNGLKM CEPCGGLCPK ACEGTGSGSR FQTVDSSNID GFVNCTKILG NLDFLITGLN GDPWHKIPAL DPEKLNVFRT VREITGYLNI QSWPPHMHNF SVFSNLTTIG GRSLYNRGFS LLIMKNLNVT SLGFRSLKEI SAGRIYISAN RQLCYHHSLN WTKVLRGPTE ERLDIKHNRP RRDCVAEGKV CDPLCSSGGC WGPGPGQCLS CRNYSRGGVC VTHCNFLNGE PREFAHEAEC FSCHPECQPM EGTATCNGSG SDTCAQCAHF RDGPHCVSSC PHGVLGAKGP IYKYPDVQNE CRPCHENCTQ GCKGPELQDC LGQTLVLIGK THLTRSPKSC DKTHTCPPCP APELLGGPSV FLFPPKPKDT LMISRTPEVT CVVVDVSHED PEVKFNWYVD GVEVHNAKTK PREEQYNSTY RVVSVLTVLH QDWLNGKEYK CKVSNKALPA PIEKTISKAK GQPREPQVYT LPPSRDELTK NQVSLTCLVK GFYPSDIAVE WESNGQPENN YKTTPPVLDS DGSFFLYSKL TVDKSRWQQG NVFSCSVMHE ALHNHYTQKS LSLSPGKHHH HHH | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ErbB3 protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |