missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ERVK-6 Protein
A DNA sequence encoding the Homo sapiens (Human) ERVK-6, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7708-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
ERVK-6 Protein | |
Research Use Only | |
ERVK-6 | |
Recombinant Protein | |
E. coli | |
LPMPAGAAAA NYTYWAYVPF PPLIRAVTWM DNPTEVYVND SVWVPGPIDD RCPAKPEEEG MMINISIGYH YPPICLGRAP GCLMPAVQNW LVEVPTVSPI CRFTYHMVSG MSLRPRVNYL QDFSYQRSLK FRPKGKPCPK EIPKESKNTE VLVWEECVAN SAVILQNNEF GTIIDWAPRG QFYHNCSGQT QSCPSAQVSP AVDSDLTESL DKHKHKKLQS FYPWEWGEKG ISTPRPKIVS PVSGPEHPEL WRLTVASHHI RIWSGNQTLE TRDRKPFYTI DLNSSLTVPL QSCVKPPYML VVGNIVIKPD SQTITCENCR LLTCIDSTFN WQHRILLVRA REGVWIPVSM DRPWEASPSV HILTEVLKGV LNRSKRFIFT LIAVIMGLIA VTATAAVAGV ALHSSVQSVN FVNDWQKNST RLWNSQSSID QKLANQINDL RQTVIWMGDR LMSLEHRFQL QCDWNTSDFC ITPQIYNESE HHWDMVRRHL QGREDNLTLD ISKLKEQIFE ASKAHLNLVP GTEAIAGVAD GLANLNPVTW VKT | |
Greater than 90% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |