missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FAM96B Protein
A DNA sequence encoding the Homo sapiens (Human) FAM96B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8299-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_057146.1 | |
FAM96B Protein | |
Research Use Only | |
FAM96B | |
Recombinant Protein | |
E. coli | |
VGGGGVGGGL LENANPLIYQ RSGERPVTAG EEDEQVPDSI DAREIFDLIR SINDPEHPLT LEELNVVEQV RVQVSDPEST VAVAFTPTIP HCSMATLIGL SIKVKLLRSL PQRFKMDVHI TPGTHASEHA VNKQLADKER VAAALENTHL LEVVNQCLSA RS | |
Greater than 80% as determined by SDS-PAGE. |
51647 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |