missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fructose-2,6-bisphosphatase TIGAR Protein
A DNA sequence encoding the Human Fructose-2,6-bisphosphatase TIGAR, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10157-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_065108.1 | |
Fructose-2,6-bisphosphatase TIGAR Protein | |
Research Use Only | |
TIGAR | |
The biological activity determined by its ability to protect U2OS cells from apoptosis induced by hydrogen peroxide is in a concentration range of 0.1-5.0 ug/ml, after pretreating with rHuTIGAR-TAT for 4 hours. | |
Human | |
TAT | |
0.2 m filtered 30 % Acetonitrile, 0.1% TFA, lyophilized |
57103 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MARFALTVVR HGETRFNKEK IIQGQGVDEP LSETGFKQAA AAGIFLNNVK FTHAFSSDL MRTKQTMHGI LERSKFCKDM TVKYDSRLRE RKYGVVEGKA LSELRAMAK AAREECPVFT PPGGETLDQV KMRGIDFFEF LCQLILKEAD QKEQFSQGS PSNCLETSLA EIFPLGKNHS SKVNSDSGIP GLAASVLVVS HGAYMRSLF DYFLTDLKCS LPATLSRSEL MSVTPNTGMS LFIINFEEGR EVKPTVQCI CMNLQDHLNG LTETRGGYGR KKRRQ | |
Greater than 96% as determined by SDS-PAGE and HPLC. |