missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fructose-bisphosphate aldolase A Protein
A DNA sequence encoding the Homo sapiens (Human) Fructose-bisphosphate aldolase A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8303-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Spécification
NP_000025.1 | |
Fructose-bisphosphate aldolase A Protein | |
Research Use Only | |
ALDOA | |
Recombinant Protein | |
E. coli | |
PYQYPALTPE QKKELSDIAH RIVAPGKGIL AADESTGSIA KRLQSIGTEN TEENRRFYRQ LLLTADDRVN PCIGGVILFH ETLYQKADDG RPFPQVIKSK GGVVGIKVDK GVVPLAGTNG ETTTQGLDGL SERCAQYKKD GADFAKWRCV LKIGEHTPSA LAIMENANVL ARYASICQQN GIVPIVEPEI LPDGDHDLKR CQYVTEKVLA AVYKALSDHH IYLEGTLLKP NMVTPGHACT QKFSHEEIAM ATVTALRRTV PPAVTGITFL SGGQSEEEAS INLNAINKCP LLKPWALTFS YGRALQASAL KAWGGKKENL KAAQEEYVKR ALANSLACQG KYTPSGQAGA AASESLFVSN HAY | |
Greater than 90% as determined by SDS-PAGE. |
226 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |