missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDF-8 / Myostatin / MSTN Protein
Recombinant Protein
Marca: enQuireBio™ QP5329-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_005250.1 | |
GDF-8 / Myostatin / MSTN Protein | |
10 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
MPC-11 cell cytotoxicity significantly induced by less than 50 ng/ml which corresponds to an activity of at least 2.0 x 10^4 units/mg. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
2660 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
MSTN | |
Recombinant Protein | |
E. coli | |
DFGLDCDEHS TESRCCRYPL TVDFEAFGWD WIIAPKRYKA NYCSGECEFV FLQKYPHTHL VHQANPRGSA GPCCTPTKMS PINMLYFNGK EQIIYGKIPA MVVDRCGCS |