missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDF15 D Protein
A cDNA sequence encoding the GDF15 D was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11949-20ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
GDF15 D Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MARNGDDCPL GPGRCCRLHT VRASLEDLGW ADWVLSPREV QVTMCIGACP SQFRAANMHA QIKTSLHRLK PDTVPAPCCV PASYNPMVLI QKTDTGVSLQ TYDDLLAKDC HCI | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
GDF15 D-variant is lyophilized without additives. |