missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GGH Protein
A cDNA sequence encoding the GGH was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11960-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
8836 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MRPHGDTAKK PIIGILMQKC RNKVMKNYGR YYIAASYVKY LESAGARVVP VRLDLTEKDY EILFKSINGI LFPGGSVDLR RSDYAKVAKI FYNLSIQSFD DGDYFPVWGT CLGFEELSLL ISGECLLTAT DTVDVAMPLN FTGGQLHSRM FQNFPTELLL SLAVEPLTAN FHKWSLSVKN FTMNEKLKKF FNVLTTNTDG KIEFISTMEG YKYPVYGVQW HPEKAPYEWK NLDGISHAPN AVKTAFYLAE FFVNEARKNN HHFKSESEEE KALIYQFSPI YTGNISSFQQ CYIFD | |
Greater than 95% as determined by SDS-PAGE. |
GGH Protein | |
Research Use Only | |
GGH | |
Human | |
His | |
The GGH solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 10% glycerol. |