missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HDHD3 Protein
A cDNA sequence encoding the HDHD3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12210-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
81932 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMAHR LQIRLLTWDV KDTLLRLRHP LGEAYATKAR AHGLEVEPSA LEQGFRQAYR AQSHSFPNYG LSHGLTSRQW WLDVVLQTFH LAGVQDAQAV APIAEQLYKD FSHPCTWQVL DGAEDTLREC RTRGLRLAVI SNFDRRLEGI LEGLGLREHF DFVLTSEAAG WPKPDPRIFQ EALRLAHMEP VVAAHVGDNY LCDYQGPRAV GMHSFLVVGP QALDPVVRDS VPKEHILPSL AHLLPALDCL EGSTPGL | |
Greater than 95.0% as determined by SDS-PAGE. |
HDHD3 Protein | |
Research Use Only | |
HDHD3 | |
Human | |
His | |
The HDHD3 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |