missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HLF Protein
A cDNA sequence encoding the HLF was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12278-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
HLF Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEKMSRPLPL NPTFIPPPYG VLRSLLENPL KLPLHHEDAF SKDKDKEKKL DDESNSPTVP QSAFLGPTLW DKTLPYDGDT FQLEYMDLEE FLSENGIPPS PSQHDHSPHP PGLQPASSAA PSVMDLSSRA SAPLHPGIPS PNCMQSPIRP GQLLPANRNT PSPIDPDTIQ VPVGYEPDPA DLALSSIPGQ EMFDPRKRKF SEEELKPQPM IKKARKVFIP DDLKDDKYWA RRRKNNMAAK RSRDARRLKE NQIAIRASFL EKENSALRQE VADLRKELGK CKNILAKYEA RHGPL | |
Greater than 85.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HLF protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 40% glycerol, 0.2M NaCl and 0.1mM PMSF. |