missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ICAM2 Protein

A cDNA sequence encoding the ICAM2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12363-2ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15950179

  • 120.00€ / 2µg

Spedizione stimata: 03-02-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Specifica

Specifica

ICAM2 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The ICAM2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSSDEKVFE VHVRPKKLAV EPKGSLEVNC STTCNQPEVG GLETSLDKIL LDEQAQWKHY LVSNISHDTV LQCHFTCSGK QESMNSNVSV YQPPRQVILT LQPTLVAVGK SFTIECRVPT VEPLDSLTLF LFRGNETLHY ETFGKAAPAP QEATATFNST ADREDGHRNF SCLAVLDLMS RGGNIFHKHS APKMLEIYEP VSDSQ
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali