missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFN-alpha / IFNA1 / IFN Protein
A DNA sequence encoding the Human IFN-alpha / IFNA1 / IFN, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10403-ec-250ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_008831.3 | |
IFN-alpha / IFNA1 / IFN Protein | |
Research Use Only | |
IFNA1 | |
The specific activity determined by an anti-viral assay is no less than 1.0 M 108 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, containing 3 % Mannitol, 5 % Trehalose, 0.05 % Tween-80, lyophilized |
3439 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MCDLPETHSL DNRRTLMLLA QMSRISPSSC LMDRHDFGFP QEEFDGNQFQ KAPAISVLHE LIQQIFNLFT TKDSSAAWDE DLLDKFCTEL YQQLNDLEAC VMQEERVGET PLMNADSILA VKKYFRRITL YLTEKKYSPC AWEVVRAEIM RSLSLSTNLQ ERLRRKE | |
Greater than 97% as determined by SDS-PAGE and HPLC. |