missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IGFBP3 Protein
A DNA sequence encoding the Human IGFBP3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10386-ec-25ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000589.2 | |
IGFBP3 Protein | |
Research Use Only | |
IGFBP3 | |
The ED50 as determined by inhibiting IGF-II induced proliferation of serum free human MCF-7 cells is less than 200 ng/ml, corresponding to a specific activity of > 5.0 M 103 IU/mg in the presence of 15 ng/ml of rHuIGF-II. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
3486 | |
25 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
GASSGGLGPV VRCEPCDARA LAQCAPPPAV CAELVREPGC GCCLTCALSE GQPCGIYTER CGSGLRCQPS PDEARPLQAL LDGRGLCVNA SAVSRLRAYL LPAPPAPGNA SESEEDRSAG EVESPSVSST HRVSDPKFHP LHSKIIIIKK GHAKDSQRYK VDYESQSTDT QNFSSESKRE TEYGPCRREM EDTLNHLKFL NVLSPRGVHI PNCDKKGFYK KKQCRPSKGR KRGFCWCVDK YGQPLPGYTT KGKEDVHCYS MQSK | |
Greater than 98% as determined by SDS-PAGE and HPLC. |