missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL 28A Protein
A cDNA sequence encoding the IL 28A was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10707-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
282616 | |
Greater than 95% as obsereved by SDS-PAGE. | |
Research Use Only | |
IFNL2 | |
Recombinant Protein | |
HEK 293 | |
MKLDMTGDCT PVLVLMAAVL TVTGAVPVAR LHGALPDARG CHIAQFKSLS PQELQAFKRA KDALEESLLL KDCRCHSRLF PRTWDLRQLQ VRERPMALEA ELALTLKVLE ATADTDPALV DVLDQPLHTL HHILSQFRAC IQPQPTAGPR TRGRLHHWLY RLQEAPKKES PGCLEASVTF NLFRLLTRDL NCVASGDLCV |
IL 28A Protein | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The specific activity was determined by the dose dependent protection of the cytopathic effect on A549 cells (human lung adenocarcinoma epithelial cell line) that were challenged with encephalomyocarditis (EMC) virus and is typically 0.5-5ng/ml. | |
Human | |
Untagged | |
The IL28A was lyophilized from 1 mg/ml in 1xPBS. |