missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL17 / IL17A Protein
A DNA sequence encoding the Human IL17 / IL17A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10323-ec-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
IL17 / IL17A Protein | |
Research Use Only | |
IL17A | |
The ED50 as determined by inducing IL-6 secretion of murine NIH/3T3 cells is less than 7.5 ng/ml, corresponding to a specific activity of > 1.3 M 105 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
GITIPRNPGC PNSEDKNFPR TVMVNLNIHN RNTNTNPKRS SDYYNRSTSP WNLHRNEDPE RYPSVIWEAK CRHLGCINAD GNVDYHMNSV PIQQEILVLR REPPHCPNSF RLEKILVSVG CTCVTPIVHH VA | |
Greater than 95% as determined by SDS-PAGE and HPLC. |