missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Interleukin-32 / IL-32 Protein
Recombinant Protein
Marca: enQuireBio™ QP5209-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001012651.1 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MCFPKVLSDD MKKLKARMHQ AIERFYDKMQ NAESGRGQVM SSLAELEDDF KEGYLETVAA YYEEQHPELT PLLEKERDGL RCRGNRSPVP DVEDPATEEP GESFCDKSYG APRGDKEELT PQKCSEPQSS K | |
Greater than 95% as determined by SDS-PAGE. |
Interleukin-32 / IL-32 Protein | |
Research Use Only | |
IL32 | |
Human | |
Untagged | |
50 mM sodium phosphate, pH 7.5 |