missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Interleukin enhancer-binding factor 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Interleukin enhancer-binding factor 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8309-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001254738.1 | |
Interleukin enhancer-binding factor 2 Protein | |
Research Use Only | |
ILF2 | |
Recombinant Protein | |
E. coli | |
MRGDRGRGRG GRFGSRGGPG GGFRPFVPHI PFDFYLCEMA FPRVKPAPDE TSFSEALLKR NQDLAPNSAE QASILSLVTK INNVIDNLIV APGTFEVQIE EVRQVGSYKK GTMTTGHNVA DLVVILKILP TLEAVAALGN KVVESLRAQD PSEVLTMLTN ETGFEISSSD ATVKILITTV PPNLRKLDPE LHLDIKVLQS ALAAIRHARW FEENASQSTV KVLIRLLKDL RIRFPGFEPL TPWILDLLGH YAVMNNPTRQ PLALNVAYRR CLQILAAGLF LPGSVGITDP CESGNFRVHT VMTLEQQDMV CYTAQTLVRI LSHGGFRKIL GQEGDASYLA SEISTWDGVI VTPSEKAYEK PPEKKEGEEE EENTEEPPQG EEEESMETQE | |
Greater than 90% as determined by SDS-PAGE. |
3608 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |