missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IP-10 / CXCL10 Protein
Recombinant Protein
Marca: enQuireBio™ QP1011-5 ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001556.2 | |
IP-10 / CXCL10 Protein | |
Research Use Only | |
CXCL10 | |
Recombinant Protein | |
E. coli | |
VPLSRTVRCT CISISNQPVN PRSLEKLEII PASQFCPRVE IIATMKKKGE KRCLNPESKA IKNLLKAVSK ERSKRSP | |
Greater than 95% as determined by SDS-PAGE. |
3627 | |
5 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human IP-10 / CXCL10 induces Human T cell chemotaxis (significanty different from control at Human IP-10 / CXCL10 concentrations greater than 5 ng/ml). | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |