missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IRF 2 Protein
A cDNA sequence encoding the IRF 2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12457-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
3660 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPVERMRMRP WLEEQINSNT IPGLKWLNKE KKIFQIPWMHAARHGWDVEK DAPLFRNWAI HTGKHQPGVD KPDPKTWKAN FRCAMNSLPD IEEVKDKSIKKGNNAFRVYR MLP | |
Greater than 95.0% as determined by SDS-PAGE. |
IRF 2 Protein | |
Research Use Only | |
IRF2 | |
Human | |
His | |
1 mg/ml in 20mM Tris pH 8.0, 1mM DTT and 10% Glycerol. |