missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Keratin, type II cytoskeletal 6A Protein
A DNA sequence encoding the Homo sapiens (Human) Keratin, type II cytoskeletal 6A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6282-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_005545.1 | |
Keratin, type II cytoskeletal 6A Protein | |
Research Use Only | |
KRT6A | |
Recombinant Protein | |
E. coli | |
ASTSTTIRSH SSSRRGFSAN SARLPGVSRS GFSSVSVSRS RGSGGLGGAC GGAGFGSRSL YGLGGSKRIS IGGGSCAISG GYGSRAGGSY GFGGAGSGFG FGGGAGIGFG LGGGAGLAGG FGGPGFPVCP PGGIQEVTVN QSLLTPLNLQ IDPTIQRVRA EEREQIKTLN NKFASFIDKV RFLEQQNKVL ETKWTLLQEQ GTKTVRQNLE PLFEQYINNL RRQLDSIVGE RGRLDSELRG MQDLVEDFKN KYEDEINKRT AAENEFVTLK KDVDAAYMNK VELQAKADTL TDEINFLRAL YDAELSQMQT HISDTSVVLS MDNNRNLDLD SIIAEVKAQY EEIAQRSRAE AESWYQTKYE ELQVTAGRHG DDLRNTKQEI AEINRMIQRL RSEIDHVKKQ CANLQAAIAD AEQRGEMALK DAKNKLEGLE DALQKAKQDL ARLLKEYQEL MNVKLALDVE IATYRKLLEG EECRLNGEGV GQVNISVVQS TVSSGYGGAS GVGSGLGLGG GSSYSYGSGL GVGGGFSSSS GRAIGGGLSS VGGGSSTIKY TTTSSSSRKS YKH | |
Greater than 90% as determined by SDS-PAGE. |
3853 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |