missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Keratin, type II cytoskeletal 6A Protein

A DNA sequence encoding the Homo sapiens (Human) Keratin, type II cytoskeletal 6A, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6282-ec-1mg

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16012170

  • 2385.00€ / 1mg
Spedizione stimata: 27-06-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Specifica

Specifica

NP_005545.1
Keratin, type II cytoskeletal 6A Protein
Research Use Only
KRT6A
Recombinant Protein
E. coli
ASTSTTIRSH SSSRRGFSAN SARLPGVSRS GFSSVSVSRS RGSGGLGGAC GGAGFGSRSL YGLGGSKRIS IGGGSCAISG GYGSRAGGSY GFGGAGSGFG FGGGAGIGFG LGGGAGLAGG FGGPGFPVCP PGGIQEVTVN QSLLTPLNLQ IDPTIQRVRA EEREQIKTLN NKFASFIDKV RFLEQQNKVL ETKWTLLQEQ GTKTVRQNLE PLFEQYINNL RRQLDSIVGE RGRLDSELRG MQDLVEDFKN KYEDEINKRT AAENEFVTLK KDVDAAYMNK VELQAKADTL TDEINFLRAL YDAELSQMQT HISDTSVVLS MDNNRNLDLD SIIAEVKAQY EEIAQRSRAE AESWYQTKYE ELQVTAGRHG DDLRNTKQEI AEINRMIQRL RSEIDHVKKQ CANLQAAIAD AEQRGEMALK DAKNKLEGLE DALQKAKQDL ARLLKEYQEL MNVKLALDVE IATYRKLLEG EECRLNGEGV GQVNISVVQS TVSSGYGGAS GVGSGLGLGG GSSYSYGSGL GVGGGFSSSS GRAIGGGLSS VGGGSSTIKY TTTSSSSRKS YKH
Greater than 90% as determined by SDS-PAGE.
3853
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali