missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Kynurenine 3-monooxygenase Protein
A DNA sequence encoding the Homo sapiens (Human) Kynurenine 3-monooxygenase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6271-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_003670.2 | |
Kynurenine 3-monooxygenase Protein | |
Research Use Only | |
KMO | |
Recombinant Protein | |
E. coli | |
MDSSVIQRKK VAVIGGGLVG SLQACFLAKR NFQIDVYEAR EDTRVATFTR GRSINLALSH RGRQALKAVG LEDQIVSQGI PMRARMIHSL SGKKSAIPYG TKSQYILSVS RENLNKDLLT AAEKYPNVKM HFNHRLLKCN PEEGMITVLG SDKVPKDVTC DLIVGCDGAY STVRSHLMKK PRFDYSQQYI PHGYMELTIP PKNGDYAMEP NYLHIWPRNT FMMIALPNMN KSFTCTLFMP FEEFEKLLTS NDVVDFFQKY FPDAIPLIGE KLLVQDFFLL PAQPMISVKC SSFHFKSHCV LLGDAAHAIV PFFGQGMNAG FEDCLVFDEL MDKFSNDLSL CLPVFSRLRI PDDHAISDLS MYNYIEMRAH VNSSWFIFQK NMERFLHAIM PSTFIPLYTM VTFSRIRYHE AVQRWHWQKK VINKGLFFLG SLIAISSTYL LIHYMSPRSF LRLRRPWNWI AHFRNTTCFP AKAVDSLEQI SNLISR | |
Greater than 90% as determined by SDS-PAGE. |
8564 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |