missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Lamin-A Protein

A cDNA sequence encoding the Lamin-A was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12528-2ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15950719

  • 137.00€ / 2µg
Spedizione stimata: 27-06-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Specifica

Specifica

4000
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
METPSQRRAT RSGAQASSTP LSPTRITRLQ EKEDLQELND RLAVYIDRVR SLETENAGLR LRITESEEVV SREVSGIKAA YEAELGDARK TLDSVAKERA RLQLELSKVR EEFKELKARN TKKEGDLIAA QARLKDLEAL LNSKEAALST ALSEKRTLEG ELHDLRGQVA KLEAALGEAK KQLQDEMLRR VDAENRLQTM KEELDFQKNI YSEELRETKR RHETRLVEID NGKQREFESR LADALQELRA QHEDQVEQYK KELEKTYSAK LDNARQSAER NSNLVGAAHE ELQQSRIRID SLSAQLSQLQ KQLAAKEAKL RDLEDSLARE RDTSRRLLAE KEREMAEMRA RMQQQLDEYQ ELLDIKLALD MEIHAYRKLL EGEEERLRLS PSPTSQRSRG RASSHSSQTQ GGGSVTKKRK LESTESRSSF SQHARTSGRV AVEEVDEEGK FVRLRNKSNE DQSMGNWQIK RQNGDDPLLT YRFPPKFTLK AGQVVTIWAA GAGATHSPPT DLVWKAQNTW GCGNSLRTAL INSTGEEVAM RKLVRSVTVV EDDEDEDGDD LLHHHHGSHC SSSGDPAEYN LRSRTVLCGT CGQPADKASA SGSGAQVGGP ISSGSSASSV TVTRSYRSVG GSGGGSFGDN LVTRSYLLGN SSPRTQSPQN CSIM
Greater than 90.0% as determined by SDS-PAGE.
Lamin-A Protein
Research Use Only
LMNA
Human
Untagged
The Lamin-A Protein solution (0.9 mg/ml) contains 20mM phosphate buffer pH 7.0, 500mM NaCl, 1mM DTT, 1.5mM EDTA and 20% (v/v) Glycerol.
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali