missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Laminin subunit alpha-5 Protein
A DNA sequence encoding the Homo sapiens (Human) Laminin subunit alpha-5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6287-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_005551.3 | |
Laminin subunit alpha-5 Protein | |
Research Use Only | |
LAMA5 | |
Recombinant Protein | |
E. coli | |
FVAQMEGLGT RLRAQSRQRS RPGRWHKVSV RWEKNRILLV TDGARAWSQE GPHRQHQGAE HPQPHTLFVG GLPASSHSSK LPVTVGFSGC VKRLRLHGRP LGAPTRMAGV TPCILGPLEA GLFFPGSGGV ITLDLPGATL PDVGLELEVR PLAVTGLIFH LGQARTPPYL QLQVTEKQVL LRADDGAGEF STSVTRPSVL CDGQWHRLAV MKSGNVLRLE VDAQSNHTVG PLLAAAAGAP APLYLGGLPE PMAVQPWPPA YCGCMRRLAV NRSPVAMTRS VEVHGAVGAS GC | |
Greater than 90% as determined by SDS-PAGE. |
3911 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |