missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LGALS8 Protein
A cDNA sequence encoding the LGALS8 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10770-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
LGALS8 Protein | |
Research Use Only | |
The ED50 for this effect is 5–10ug/ml. Measured by its ability to agglutinate human red blood cells corresponding to a specific activity of 100-200IU/mg. | |
Human | |
His | |
The Galectin-8 His tag 0.5 mg/ml protein solution contains 20mM Tris-HCl pH 8.0, 0.1M NaCl, 10% glycerol and 1mM DTT. |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MMLSLNNLQN IIYNPVIPFV GTIPDQLDPG TLIVIRGHVP SDADRFQVDL QNGSSMKPRA DVAFHFNPRF KRAGCIVCNT LINEKWGREE ITYDTPFKRE KSFEIVIMVL KDKFQVAVNG KHTLLYGHRI GPEKIDTLGI YGKVNIHSIG FSFSSDLQST QASSLELTEI SRENVPKSGT PQLRLPFAAR LNTPMGPGRT VVVKGEVNAN AKSFNVDLLA GKSKDIALHL NPRLNIKAFV RNSFLQESWG EEERNITSFP FSPGMYFEMI IYCDVREFKV AVNGVHSLEY KHRFKELSSI DTLEINGDIH LLEVRSW | |
Greater than 90.0% as determined by SDS-PAGE. |