missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LSM3 Protein
A cDNA sequence encoding the LSM3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12592-20ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
LSM3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMADDVDQ QQTTNTVEEP LDLIRLSLDE RIYVKMRNDR ELRGRLHAYD QHLNMILGDV EETVTTIEID EETYEEIYKS TKRNIPMLFV RGDGVVLVAP PLRVG | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The LSM3 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM Nacl, 2mM DTT and 20% glycerol. |