missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Lysyl oxidase homolog 4 Protein
A DNA sequence encoding the Homo sapiens (Human) Lysyl oxidase homolog 4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7943-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_115587.6 | |
Lysyl oxidase homolog 4 Protein | |
Research Use Only | |
Loxl4 | |
Recombinant Protein | |
E. coli | |
QSLGTTKLRL VGPESKPEEG RLEVLHQGQW GTVCDDNFAI QEATVACRQL GFEAALTWAH SAKYGQGEGP IWLDNVRCVG TESSLDQCGS NGWGVSDCSH SEDVGVICHP RRHRGYLSET VSNALGPQGR RLEEVRLKPI LASAKQHSPV TEGAVEVKYE GHWRQVCDQG WTMNNSRVVC GMLGFPSEVP VDSHYYRKVW DLKMRDPKSR LKSLTNKNSF WIHQVTCLGT EPHMANCQVQ VAPARGKLRP ACPGGMHAVV SCVAGPHFRP PKTKPQRKGS WAEEPRVRLR SGAQVGEGRV EVLMNRQWGT VCDHRWNLIS ASVVCRQLGF GSAREALFGA RLGQGLGPIH LSEVRCRGYE RTLSDCPALE GSQNGCQHEN DAAVRCNVPN MGFQNQVRLA GGRIPEEGLL EVQVEVNGVP RWGSVCSENW GLTEAMVACR QLGLGFAIHA YKETWFWSGT PRAQEVVMSG VRCSGTELAL QQCQRHGPVH CSHGGGRFLA GVSCMDSAPD LVMNAQLVQE TAYLEDRPLS QLYCAHEENC LSKSADHMDW PYGYRRLLRF STQIYNLGRT DFRPKTGRDS WVWHQCHRHY HSIEVFTHYD LLTLNGSKVA EGHKASFCLE DTNCPTGLQR RYACANFGEQ GVTVGCWDTY RHDIDCQWVD ITDVGPGNYI FQVIVNPHYE VAESDFSNNM LQCRCKYDGH RVWLHNCHTG NSYPANAELS LEQEQRLRNN LI | |
Greater than 90% as determined by SDS-PAGE. |
84171 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |