missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAGEA5 Protein
A cDNA sequence encoding the MAGEA5 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12624-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
MAGEA5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSLEQK SQHCKPEEGL DTQEEALGLV GVQAATTEEQ EAVSSSSPLV PGTLGEVPAA GSPGPLKSPQ GASAIPTAID FTLWRQSIKG SSNQEEEGPS TSPDPESVFR AALSKKVADL IHFLLLKY | |
Greater than 85% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MAGEA5 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1mM PMSF and 20% glycerol. |