missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Mammaglobin-A Protein
A DNA sequence encoding the Homo sapiens (Human) Mammaglobin-A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7590-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002402.1 | |
Mammaglobin-A Protein | |
Research Use Only | |
SCGB2A2 | |
Recombinant Protein | |
E. coli | |
GSGCPLLENV ISKTINPQVS KTEYKELLQE FIDDNATTNA IDELKECFLN QTDETLSNVE VFMQLIYDSS LCDLF | |
Greater than 80% as determined by SDS-PAGE. |
4250 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |