missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MG Protein
A cDNA sequence encoding the MG was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10785-10mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
MG Protein | |
Research Use Only | |
100 IU/mg of FSH and 100 IU/mg LH. | |
Human | |
Untagged |
10 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Human | |
The Human MG was lyophilized from a concentrated (1 mg/ml) solution with no additives. |