missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MIF / GLIF Protein
Recombinant Protein
Marca: enQuireBio™ QP5276-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002406.1 | |
MIF / GLIF Protein | |
Research Use Only | |
MIF | |
Recombinant Protein | |
E. coli | |
MPMFIVNTNV PRASVPDGFL SELTQQLAQA TGKPPQYIAV HVVPDQLMAF GGSSEPCALC SLHSIGKIGG AQNRSYSKLL CGLLAERLRI SPDRVYINYY DMNAANVGWN NSTFA | |
Greater than 95% as determined by SDS-PAGE. |
4282 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human MIF / GLIF induces IL-8 production in human PBMCs. Activity was further confirmed by measuring chemotaxisof U937 cells via agarose microdroplet assay. | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |