missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MMP28 Protein
A cDNA sequence encoding the MMP28 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12710-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
MMP28 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSFAKQGNK WYKQHLSYRL VNWPEHLPEP AVRGAVRAAF QLWSNVSALE FWEAPATGPA DIRLTFFQGD HNDGLGNAFD GPGGALAHAF LPRRGEAHFD QDERWSLSRR RGRNLFVVLA HEIGHTLGLT HSPAPRALMA PYYKRLGRDA LLSWDDVLAV QSLYGKPLGG SVAVQLPGKL FTDFETWDSY SPQGRRPETQ GPKYCHSSFD AITVDRQQQL YIFKGSHFWE VAADGNVSEP RPLQERWVGL PPNIEAAAVS LNDGDFYFFK GGRCWRFRGP KPVWGLPQLC RAGGLPRHPD AALFFPPLRR LILFKGARYY VLARGGLQVE PYYPRSLQDW GGIPEEVSGA LPRPDGSIIF FRDDRYWRLD QAKLQATTSG RWATELPWMG CWHANSGSAL F | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MMP28 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |