missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myomegalin Protein
A DNA sequence encoding the Homo sapiens (Human) Myomegalin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7688-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001002810.1 | |
Myomegalin Protein | |
Research Use Only | |
PDE4DIP | |
Recombinant Protein | |
E. coli | |
MKGTDSGSCC RRRCDFGCCC RASRRAHYTP YRSGDATRTP QSPRQTPSRE RRRPEPAGSW AAAAEEEEAA AAATPWMRDY FAEDDGEMVP RTSHTAAFLS DTKDRGPPVQ SQIWRSGEKV PFVQTYSLRA FEKPPQVQTQ ALRDFEKHLN DLKKENFSLK LRIYFLEERM QQKYEASRED IYKRNIELKV EVESLKRELQ DKKQHLDKTW ADVENLNSQN EAELRRQFEE RQQETEHVYE LLENKIQLLQ EESRLAKNEA ARMAALVEAE KECNLELSEK LKGVTKNWED VPGDQVKPDQ YTEALAQRDK | |
Greater than 80% as determined by SDS-PAGE. |
653513 | |
200 μg | |
Not determined. | |
Full Length of Isoform 8 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |