missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDUFS5 Protein
A DNA sequence encoding the Homo sapiens (Human) NDUFS5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8422-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001171908.1 | |
NDUFS5 Protein | |
Research Use Only | |
NDUFS5 | |
Recombinant Protein | |
E. coli | |
PFLDIQKRFG LNIDRWLTIQ SGEQPYKMAG RCHAFEKEWI ECAHGIGYTR AEKECKIEYD DFVECLLRQK TMRRAGTIRK QRDKLIKEGK YTPPPHHIGK GEPRP | |
Greater than 90% as determined by SDS-PAGE. |
4725 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |