missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NEUROG3 Protein
A cDNA sequence encoding the NEUROG3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12832-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
50674 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MTPQPSGAPT VQVTRETERS FPRASEDEVT CPTSAPPSPT RTRGNCAEAE EGGCRGAPRK LRARRGGRSR PKSELALSKQ RRSRRKKAND RERNRMHNLN SALDALRGVL PTFPDDAKLT KIETLRFAHN YIWALTQTLR IADHSLYALE PPAPHCGELG SPGGSPGDWG SLYSPVSQAG SLSPAASLEE RPGLLGATSS ACLSPGSLAF SDFL | |
Greater than 85% as determined by SDS-PAGE. |
NEUROG3 Protein | |
Research Use Only | |
NEUROG3 | |
Human | |
His | |
The NEUROG3 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |