missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Neuropeptide FF receptor 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Neuropeptide FF receptor 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9310-ye-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001138228.1 | |
Neuropeptide FF receptor 2 Protein | |
Research Use Only | |
NPFFR2 | |
Recombinant Protein | |
Yeast | |
MNEKWDTNSS ENWHPIWNVN DTKHHLYSDI NITYVNYYLH QPQVAAIFII SYFLIFFLCM MGNTVVCFIV MRNKHMHTVT NLFILNLAIS DLLVGIFCMP ITLLDNIIAG WPFGNTMCKI SGLVQGISVA ASVFTLVAIA VDRFQCVVYP FKPKLTIKTA FVIIMIIWVL AITIMSPSAV MLHVQEEKYY RVRLNSQNKT SPVYWCREDW PNQEMRKIYT TVLFANIYLA PLSLIVIMYG RIGISLFRAA VPHTGRKNQE QWHVVSRKKQ KIIKMLLIVA LLFILSWLPL WTLMMLSDYA DLSPNELQII NIYIYPFAHW LAFGNSSVNP IIYGFFNENF RRGFQEAFQL QLCQKRAKPM EAYALKAKSH VLINTSNQLV QESTFQNPHG ETLLYRKSAE KPQQELVMEE LKETTNSSEI | |
Greater than 90% as determined by SDS-PAGE. |
10886 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |