missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NFASC / Neurofascin Protein
A DNA sequence encoding the Homo sapiens (Human) NFASC / Neurofascin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6416-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001005388.2 | |
NFASC / Neurofascin Protein | |
Research Use Only | |
NFASC | |
Recombinant Protein | |
E. coli | |
KRSRGGKYPV REKKDVPLGP EDPKEEDGSF DYSDEDNKPL QGSQTSLDGT IKQQESDDSL VDYGEGGEGQ FNEDGSFIGQ YTVKKDKEET EGNESSEATS PVNAIYSLA | |
Greater than 90% as determined by SDS-PAGE. |
23114 | |
50 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |