missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NMM Protein
A cDNA sequence encoding the NMM was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10792-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NMM Protein | |
Research Use Only | |
Assayed for phosphorilation by MLCK. | |
Human | |
Untagged | |
NMM protein at 0.2 mg/ml in 20mM HEPES-KOH, pH 7, 50mM NaCl, 1mM EDTA and 1mM DTT. |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MSSKKAKTKT TKKRPQRATS NVFAMFDQSQ IQEFKEAFNM IDQNRDGFID KEDLHDMLAS LGKNPTDAYL DAMMNEAPGP INFTMFLTMF GEKLNGTDPE DVIRNAFACF DEEATGTIQE DYLRELLTTM GDRFTDEEVD ELYREAPIDK KGNFNYIEFT RILKHGAKDK DD | |
Greater than 95.0% as determined by SDS-PAGE. |