missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NUDC Protein
A cDNA sequence encoding the NUDC was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12897-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
10726 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGGEQEE ERFDGMLLAM AQQHEGGVQE LVNTFFSFLR RKTDFFIGGE EGMAEKLITQ TFSHHNQLAQ KTRREKRARQ EAERREKAER AARLAKEAKS ETSGPQIKEL TDEEAERLQL EIDQKKDAEN HEAQLKNGSL DSPGKQDTEE DEEEDEKDKG KLKPNLGNGA DLPNYRWTQT LSELDLAVPF CVNFRLKGKD MVVDIQRRHL RVGLKGQPAI IDGELYNEVK VEESSWLIED GKVVTVHLEK INKMEWWSRL VSSDPEINTK KINPENSKLS DLDSETRSMV EKMMYDQRQK SMGLPTSDEQ KKQEILKKFM DQHPEMDFSK AKFN | |
Greater than 90.0% as determined by SDS-PAGE. |
NUDC Protein | |
Research Use Only | |
NUDC | |
Human | |
His | |
NUDC protein solution (1 mg/ml) containing 20mM Tris-HCl (pH 8.0), 10% glycerol and 1mM DTT. |