missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PAH / PH Protein
A DNA sequence encoding the Homo sapiens (Human) PAH / PH, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6463-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000268.1 | |
PAH / PH Protein | |
Research Use Only | |
PAH | |
Recombinant Protein | |
E. coli | |
STAVLENPGL GRKLSDFGQE TSYIEDNCNQ NGAISLIFSL KEEVGALAKV LRLFEENDVN LTHIESRPSR LKKDEYEFFT HLDKRSLPAL TNIIKILRHD IGATVHELSR DKKKDTVPWF PRTIQELDRF ANQILSYGAE LDADHPGFKD PVYRARRKQF ADIAYNYRHG QPIPRVEYME EGKKTWGTVF KTLKSLYKTH ACYEYNHIFP LLEKYCGFHE DNIPQLEDVS QFLQTCTGFR LRPVAGLLSS RDFLGGLAFR VFHCTQYIRH GSKPMYTPEP DICHELLGHV PLFSDRSFAQ FSQEIGLASL GAPDEYIEKL ATIYWFTVEF GLCKQGDSIK AYGAGLLSSF GELQYCLSEK PKLLPLELEK TAIQNYTVTE FQPLYYVAES FNDAKEKVRN FAATIPRPFS VRYDPYTQRI EVLDNTQQLK ILADSINSEI GILCSALQKI K | |
Greater than 80% as determined by SDS-PAGE. |
5053 | |
10 μg | |
Not determined. | |
Full Length of Mature. Protein | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |