missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PARD6B Protein
A cDNA sequence encoding the PARD6B was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12962-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
84612 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMNRSHRH GAGSGCLGTM EVKSKFGAEF RRFSLERSKP GKFEEFYGLL QHVHKIPNVD VLVGYADIHG DLLPINNDDN YHKAVSTANP LLRIFIQKKE EADYSAFGTD TLIKKKNVLT NVLRPDNHRK KPHIVISMPQ DFRPVSSIID VDILPETHRR VRLYKYGTEK PLGFYIRDGS SVRVTPHGLE KVPGIFISRL VPGGLAQSTG LLAVNDEVLE VNGIEVSGKS LDQVTDMMIA NSRNLIITVR PANQRNNVVR NSRTSGSSGQ STDNSLLGYP QQIEPSFEPE DEDSEEDDII IEDNGVPQQI PKAVPNTESL ESLTQIELSF ESGQNGFIPS NEVSLAAIAS SSNTEFETHA PDQKLLEEDG TIITL | |
Greater than 85.0% as determined by SDS-PAGE. |
PARD6B Protein | |
Research Use Only | |
PARD6B | |
Human | |
His | |
The PARD6B solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2M Urea, 20% glycerol and 0.2M NaCl. |