missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Parvalbumin / PVALB Protein
A DNA sequence encoding the Homo sapiens (Human) Parvalbumin / PVALB, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6558-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001302461.1 | |
Parvalbumin / PVALB Protein | |
Research Use Only | |
PVALB | |
Recombinant Protein | |
E. coli | |
SMTDLLNAED IKKAVGAFSA TDSFDHKKFF QMVGLKKKSA DDVKKVFHML DKDKSGFIEE DELGFILKGF SPDARDLSAK ETKMLMAAGD KDGDGKIGVD EFSTLVAES | |
Greater than 90% as determined by SDS-PAGE. |
5816 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |